iFluor® 680-Wheat Germ Agglutinin (WGA) Conjugate
Leverandør: AAT BIOQUEST
25560
AATB25560EA 1700 NOK
AATB25560
iFluor® 680-Wheat Germ Agglutinin (WGA) Conjugate
Systemer for proteinkobling og -merking
iFluor® 680 WGA conjugate, like Alexa Fluor® 680 WGA conjugate, retains its ability to bind to sialic acid and N-acetylglucosaminyl residues, enhancing its utility in fluorescence imaging and analysis for various scientific investigations.
  • Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
NB: For research use only.