Concanavalin A, XFD594
Leverandør: AAT BIOQUEST
25575
AATB25575EA 1675 NOK
AATB25575
Concanavalin A, XFD594
Proteiner
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
  • Well-cited conjugate
  • High affinity and selectivity
  • Multiple applications
NB: For research use only.