APC maleimide, ReadiUse™, preactivated [Activated Allophycocyanin]
Leverandør: AAT BIOQUEST
2567 2568
AATB2567EA 2425 NOK
AATB2567 AATB2568
APC maleimide, ReadiUse™, preactivated [Activated Allophycocyanin]
APC maleimide
ReadiUse™ Preactivated APC Maleimide is prepared by reacting highly purified allophycocyanin (APC) with SMCC.
  • Stable for 6 months
Quality tested in-house
NB: For Research Use Only