Lumiwox™ Acridinium NHS Ester
Leverandør: AAT BIOQUEST
26000
AATB26000EA 3600 NOK
AATB26000
Lumiwox™ Acridinium NHS Ester
Assay Proteinanalyser
The acridinium NHS ester can be used to label proteins and nucleic acids.
  • Chemiluminescent labeling
  • Improved sensitivity and stronger signal
  • Tolerates hydrogen peroxide reaction
  • Suitable for various experimental surfaces
  • Multiple applications
NB: For research use only.