ATTO 488 maleimide
Leverandør: AAT BIOQUEST
2816
AATB2816EA 3625 NOK
AATB2816
ATTO 488 maleimide
ATTO 488 maleimide
ATTO 488 is a hydrophilic, rhodamine-based fluorescent dye with exceptional water solubility.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only