ATTO 647 maleimide
Leverandør: AAT BIOQUEST
2853
AATB2853EA 3625 NOK
AATB2853
ATTO 647 maleimide
ATTO 647 maleimide
ATTO 647 is a rhodamine-derived fluorescent dye optimized for applications within the red spectral region, offering spectral characteristics similar to Cy5.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only