ATTO 647N acid
Leverandør: AAT BIOQUEST
2854
AATB2854EA 9075 NOK
AATB2854
ATTO 647N acid
ATTO 647N acid
ATTO 647N is a rhodamine-based fluorescent dye optimized for use in the red spectral region, with similar spectral characteristics as Cy5.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only