Biotin hydrazide ≥95%

Leverandør: AAT BIOQUEST
Warning

3007
AATB3007EA 1425 NOK
AATB3007
Biotin hydrazide ≥95%
Biotin hydrazide

Biotin and biotin derivatives bind with high affinity to both avidins and streptavidins.

Formel: C₁₀H₁₈N₄O₂S
MDL nummer: MFCD00078532
CAS-nummer: 66640-86-6
ADR: 5,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR