5-Carboxy-4,7-dichlororhodamine 110 (5-dR110) ≥95%
Leverandør: AAT BIOQUEST
315
AATB315EA 89750 NOK
AATB315
5-Carboxy-4,7-dichlororhodamine 110 (5-dR110) ≥95%
5-Carboxy-4,7-dichlororhodamine 110 (5-dR110)
Compared to fluorescein labeling reagents such as FITC and FAM, 5-dR110 [5-carboxy-4,7-dichlororhodamine 110] reagents give more photostable and pH-independent bioconjugates that have the absorption maximum around the preferred 488 nm excitation wavelength.
  • Critical DNA Sequencing Dye
NB: For research use only.