6-Carboxy-4,7-dichlororhodamine 110 (6-dR110) ≥95%
Leverandør: AAT BIOQUEST
316
AATB316EA 89750 NOK
AATB316
6-Carboxy-4,7-dichlororhodamine 110 (6-dR110) ≥95%
6-Carboxy-4,7-dichlororhodamine 110 (6-dR110)
Compared to fluorescein labeling reagents such as FITC and FAM, 6-dR110 [6-carboxy-4,7-dichlororhodamine 110] reagents give more photostable and pH-independent bioconjugates that have the absorption maximum around the preferred 488 nm excitation wavelength.
  • Critical DNA Sequencing Dye
NB: For research use only.