5-dR110 succinimidyl ester ≥75%
Leverandør: AAT BIOQUEST
317
AATB317EA 85550 NOK
AATB317
5-dR110 succinimidyl ester ≥75%
5-dR110 succinimidyl ester
Compared to fluorescein labeling reagents such as FITC and FAM, 5-dR110 [5-carboxy-4,7-dichlororhodamine 110] reagents give more photostable and pH-independent bioconjugates that have the absorption maximum around the preferred 488 nm excitation wavelength.
  • Sanger sequencing building block
NB: For research use only.