5-Carboxyrhodamine 110 (5-CR110) single isomer ≥95%
Leverandør: AAT BIOQUEST
322
AATB322EA 5400 NOK
AATB322
5-Carboxyrhodamine 110 (5-CR110) single isomer ≥95%
5-Carboxyrhodamine 110 (5-CR110)
Compared to fluorescein labeling reagents such as FITC and FAM, CR110 reagents give more photostable and pH-independent bioconjugates that have the absorption maximum around the preferred 488 nm excitation wavelength.
  • Photostable
  • pH-independent Fluorescence
NB: For research use only.