CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
Leverandør: AAT BIOQUEST
37010 37011
AATB37010EA 1300 NOK
AATB37010 AATB37011
CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
Buffere Buffere, cellekultur
CytoWatch™ Multiplexing Buffer is a buffer optimised for the multicolor flow cytometric analysis of cells.
  • Designed for ease of use
  • Enhances multicolor flow experiments with two or more staining reagents
CytoWatch™ Multiplexing Buffer is a concentrated solution that is directly added to the staining solutions of certain fluorescent antibody conjugates or other fluorescent reagents before staining cells.
NB: For research use only.