Psoralen-PEG3-Biotin
Leverandør: AAT BIOQUEST
39050 39051
AATB39050EA 1575 NOK
AATB39050 AATB39051
Psoralen-PEG3-Biotin
Psoralen-PEG3-Biotin
Psoralen-PEG3-Biotin is a photoactivatable reagent for biotinylating DNA or RNA.
  • Stable for 24 months
Quality tested in-house
NB: For Research Use Only