Psoralen-A17-Biotin, ReadiLink™
Leverandør: AAT BIOQUEST
39052 39053
AATB39052EA 2050 NOK
AATB39052 AATB39053
Psoralen-A17-Biotin, ReadiLink™
Psoralen-A17-Biotin
ReadiLink™ Psoralen-A17-Biotin is a photoactivatable reagent for biotinylating DNA or RNA.
  • Stable for 24 months
Quality tested in-house
NB: For Research Use Only