Psoralen MOP maleimide
Leverandør: AAT BIOQUEST
39056
AATB39056EA 3125 NOK
AATB39056
Psoralen MOP maleimide
Psoralen MOP maleimide
Psoralens and their derivatives (such as 8-MOP and 4,5’8-TMP) are well known to have unique crosslinking features to DNA.
  • Stable for 24 months
Quality tested in-house
NB: For Research Use Only