Psoralen MOP azide
Leverandør: AAT BIOQUEST
39057
AATB39057EA 3125 NOK
AATB39057
Psoralen MOP azide
Psoralen MOP azide
Psoralen MOP Azide is an excellent building block for developing site-specific nucleic acid probes via the well-known click chemistry, i.e., Copper-Catalyzed Alkyne–Azide Cycloaddition (CuAAC).
  • Stable for 24 months
Quality tested in-house
NB: For Research Use Only
New Product