Psoralen TMP maleimide
Leverandør: AAT BIOQUEST
39061
AATB39061EA 3125 NOK
AATB39061
Psoralen TMP maleimide
Psoralen TMP maleimide
Psoralens and their derivatives (such as 8-TMP and 4,5’,8-TMP) are well known to have unique crosslinking features to DNA.
  • Stable for 24 months
Quality tested in-house
NB: For Research Use Only