Amplite® Tyramide (TSA) UV Channel Signal Scavenger *10X*
Leverandør: AAT BIOQUEST
44500
AATB44500EA 3175 NOK
AATB44500
Amplite® Tyramide (TSA) UV Channel Signal Scavenger *10X*
Nukleinsyre reagenser Nucleic Acid Modification and Labelling
Tyramide signal amplification (TSA) is a technique used to enhance the signal of target molecules, such as antibodies or nucleic acid probes.
  • Stable for 12 months
Quality tested in-house.
NB: For research use only.
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon