iFluor® 568, Styramide *Superior Replacement for Alexa Fluor 568 tyramide*
Leverandør: AAT BIOQUEST
45030
AATB45030EA 5400 NOK
AATB45030
iFluor® 568, Styramide *Superior Replacement for Alexa Fluor 568 tyramide*
iFluor®
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 to 50 times higher than the widely used tyramide (TSA) reagents.
  • 10 to 50X higher fluorescence
  • Improved precision and sensitivity
  • More efficient use of primary antibody
  • Enhanced water solubility
  • More photostable
NB: For research use only.