iFluor® 570 styramide [Superior Replacement for Alexa Fluor 568 tyramide]
Leverandør: AAT BIOQUEST
45031
AATB45031EA 5450 NOK
AATB45031
iFluor® 570 styramide [Superior Replacement for Alexa Fluor 568 tyramide]
iFluor® 570 styramide
iFluor® 570 Styramide is a replacement for Alexa Fluor™ 568 tyramide and similar fluorescent tyramide conjugates or TSA reagents, offering unmatched performance in sensitive imaging applications.
  • Stable for 12 months
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundance targets in cells and tissues, with fluorescence signals 10-50 times higher than tyramide (TSA) reagents.
NB: For Research Use Only