Cy7 styramide [Superior Replacement for Cy7 tyramide]
Leverandør: AAT BIOQUEST
45066
AATB45066EA 5025 NOK
AATB45066
Cy7 styramide [Superior Replacement for Cy7 tyramide]
Cy7 styramide
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only