iFluor® 430 tyramide [Superior Replacement for Opal 480]
Leverandør: AAT BIOQUEST
45096
AATB45096EA 5025 NOK
AATB45096
iFluor® 430 tyramide [Superior Replacement for Opal 480]
iFluor® 430 tyramide
iFluor® 430 tyramide is optimized to a superior replacement for Opal 480 or other spectrally similar fluorescent tyramide conjugates or TSA reagents.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only