Anti-IgG Goat Polyclonal Antibody (iFluor® 820)

Leverandør: AAT BIOQUEST
48058 48059
AATB48058EA 1625 NOK
AATB48058 AATB48059
Anti-IgG Goat Polyclonal Antibody (iFluor® 820)
Antistoffer
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography

iFluor® 820 goat anti-rabbit IgG (H+L) conjugates are ideal for various NIR imaging applications, including Western blotting, ELISA, protein arrays, tissue section imaging, and in vivo imaging.

NB: Research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 820
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG
Environmentally Preferable
Antigen-navn Immunoglobulin G
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering iFluor® 820
Reaktivitet Rabbit
Vert Goat
Isotype IgG (H, L)
Rensing Affinity chromatography
Kyssabsorpsjon No
Lagringstemp Freeze (<−15 °C); Minimise light exposure
Frakttemp. 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR