Native Horseradish Horseradish peroxidase (from Serum), Spectinomycin
Leverandør: AAT BIOQUEST
50037
AATB50037EA 9000 NOK
AATB50037
Native Horseradish Horseradish peroxidase (from Serum), Spectinomycin
Proteiner
Spectinomycin-HRP conjugate is used for developing an ELISA assay for quantifying spectinomycin.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
NB: For research use only.
Bestill nå
SPESIFIKASJONER
Protein/peptide name Horseradish peroxidase
Protein synonyms HRP
Environmentally Preferable
Protein/peptide type Native
Arter Horseradish
Kilde Serum
Konjugering Spectinomycin