Native Bovine Bovine Serum Albumin (from Serum), Spectinomycin
Leverandør: AAT BIOQUEST
50038
AATB50038EA 9000 NOK
AATB50038
Native Bovine Bovine Serum Albumin (from Serum), Spectinomycin
Proteiner
Spectinomycin-BSA conjugate is used for developing anti-spectinomycin antibodies that are typically used in an ELISA assay.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
NB: For research use only.