Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (XFD750)

Leverandør: AAT BIOQUEST
50182 50183
AATB50182EA 1800 NOK
AATB50182 AATB50183
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (XFD750)
Antistoffer
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity purified
  • Easily reconstituted

Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

NB: For research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: XFD750
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG
Environmentally Preferable
Antigen-navn Immunoglobulin G
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering XFD750
Reaktivitet Human
Vert Goat
Isotype IgG (H, L)
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Flowcytometri Yes
Kyssabsorpsjon Minimal cross reaction to human, horse, mouse, rabbit and bovine
Lagringstemp Freeze (<–15 °C); Minimise light exposure
Frakttemp. 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR