Native Horseradish Horseradish peroxidase (from Serum), Fentanyl
Leverandør: AAT BIOQUEST
50564
AATB50564EA 8600 NOK
AATB50564
Native Horseradish Horseradish peroxidase (from Serum), Fentanyl
Proteiner
Fentanyl-HRP conjugate is an essential tool for developing fentanyl assays and tests.
  • High immunogenicity
  • Useful for antibody development
  • Fentanyl test building block
NB: For research use only.
Bestill nå
SPESIFIKASJONER
Protein/peptide name Horseradish peroxidase
Protein synonyms HRP
Environmentally Preferable
Protein/peptide type Native
Arter Horseradish
Kilde Serum
Konjugering Fentanyl