Fentanyl-KLH conjugate
Leverandør: AAT BIOQUEST
50567
AATB50567EA 8225 NOK
AATB50567
Fentanyl-KLH conjugate
Fentanyl-KLH
Small compounds (drugs or peptides) are not sufficiently complex by themselves to induce an immune response or be processed in a manner that elicits production of specific antibodies.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only