Xylazine-KLH
Leverandør: AAT BIOQUEST
50621
AATB50621EA 15225 NOK
AATB50621
Xylazine-KLH
Xylazine-KLH
Xylazine-KLH conjugate is an excellent antigen for developing anti-Xylazine antibodies for ELISA-based assays and tests for detecting Xylazine.
  • Stable for 24 months
Quality tested in-house
NB: For Research Use Only