Norfentanyl-KLH conjugate
Leverandør: AAT BIOQUEST
50630
AATB50630EA 15225 NOK
AATB50630
Norfentanyl-KLH conjugate
Norfentanyl-KLH
Norfentanyl-KLH Conjugate is an excellent antigen to develop anti-norfentanyl antibodies that can be used for detecting norfentanyl substances.
  • Stable for 24 months
Quality tested in-house
NB: For Research Use Only