Statine 95

Leverandør: AAT BIOQUEST
Warning

5109
AATB5109EA 3600 NOK
AATB5109
Statine 95
Statine

(3S,4S)-Statine is a novel amino acid of pepstatin, a low molecular weight inhibitor of acid proteases.

Formel: C₈H₁₇NO₃
MW: 175.228 g/mol
MDL nummer: MFCD00037277
CAS-nummer: 49642-07-1
ADR: 5 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR