7-Hydroxycoumarin-4-acetic acid succinimidyl ester ≥90%
Leverandør: AAT BIOQUEST
553
AATB553EA 1800 NOK
AATB553
7-Hydroxycoumarin-4-acetic acid succinimidyl ester ≥90%
7-Hydroxycoumarin-4-acetic acid succinimidyl ester
7-Hydroxycoumarin-4-acetic acid is used for labeling peptides and proteins to give blue fluorescence.
  • Multiple Bioconjugation Applications
  • 90% Purity
NB: For research use only.