Amplite® Rapid Colorimetric Biotin Quantitation Kit
Leverandør: AAT BIOQUEST
5537
AATB5537EA 3625 NOK
AATB5537
Amplite® Rapid Colorimetric Biotin Quantitation Kit
Assay Proteinanalyser
Optimised to use with Nanodrop. Amplite™ colorimetric biotin quantitation kit is designed to quantify the number of biotins attached to a protein or other macromolecule with a nanodrop spectrometer.
  • Absorption assessed at 500 nm
  • The kit is best used to determine biotin concentration in the range from 2 to 16 µM
  • 12-month shelf life
It is optimised to use with Nanodrop spectrometer.
Leveringsinformasjon: Shipped with blue ice.
NB: Research use only.