Amplite® Fluorimetric Biotin Quantitation Kit
Leverandør: AAT BIOQUEST
5538
AATB5538EA 3625 NOK
AATB5538
Amplite® Fluorimetric Biotin Quantitation Kit
Assay Proteinanalyser
Designed for consistent and dependable results. Amplite® Fluorimetric Biotin Quantitation Kit uses Biotinylite™ Green, a fluorogenic biotin sensor.
  • Robust protocol
  • Consistent results
  • 12-month shelf life
This fluorescence-based assay is much more sensitive than the commonly used colorimetric HABA assay.
Leveringsinformasjon: Shipped with blue ice.
NB: Research use only.