Portelite™ Rapid Fluorimetric Biotin Quantitation Kit
Leverandør: AAT BIOQUEST
5539
AATB5539EA 3625 NOK
AATB5539
Portelite™ Rapid Fluorimetric Biotin Quantitation Kit
Assay Proteinanalyser
Optimised to use with CytoCite™ and Qubit™ fluorometers. Portelite™ rapid fluorimetric biotin quantitation kit is specifically optimised to quantify biotin and biotin conjugates with a portal fluorometer such as CytoCite™ and Qubit™ fluorometer.
  • Robust protocol
  • Consistent results
  • 12-month shelf life
This fluorescence-based assay is much more sensitive than the commonly used colorimetric HABA assay.
Leveringsinformasjon: Shipped with blue ice.
NB: Research use only.