ReadiLink™ Rapid Cy5 Antibody Labelling Kit
Leverandør: AAT BIOQUEST
5725
AATB5725EA 8050 NOK
AATB5725
ReadiLink™ Rapid Cy5 Antibody Labelling Kit
Systemer for proteinkobling og -merking
ReadiLink™ Rapid antibody labelling kits, designed for production scale, provide a convenient and efficient method for labelling large volumes of antibodies with Cy5.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
NB: Research use only.