DY547 CEP (DyLight 547 Phosphoramidite)
Leverandør: AAT BIOQUEST
6074
AATB6074EA 8150 NOK
AATB6074
DY547 CEP (DyLight 547 Phosphoramidite)
Systemer for proteinkobling og -merking
DY547 CEP (DyLight 547 phosphoramidite) is an excellent replacement to the commonly used Cy3 phosphoramidite.
  • High stability
  • Oligo deprotection can be carried out at room temperature
DY547 CEP appears to be more stable than Cy3 phosphoramidite. As Cy3 and Cy5 phosphoramidites, the deprotection of oligos containing DY547 CEP may be carried out with ammonium hydroxide at room temperature, regardless of the base protecting groups on the monomers used..
It is always prudent to use monomers with base labile protecting groups to limit the exposure time to 2 hours or less at 65 °C during deprotection.
Forpakning: Plastic vial.
NB: For research use only.