Cy5 Dye qPCR Calibration Solution *10000X*
Leverandør: AAT BIOQUEST
67033
AATB67033EA 4925 NOK
AATB67033
Cy5 Dye qPCR Calibration Solution *10000X*
Nukleinsyre reagenser qPCR/RT-qPCR Enzymes and Kits
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
  • Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
NB: For research use only.