TAMRA Dye qPCR Calibration Solution *10000X*
Leverandør: AAT BIOQUEST
67036
AATB67036EA 4925 NOK
AATB67036
TAMRA Dye qPCR Calibration Solution *10000X*
Nukleinsyre reagenser qPCR/RT-qPCR Enzymes and Kits
TAMRA dye qPCR Calibration Solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ 7 systems
  • Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
NB: For research use only.