MP650 Dye qPCR Calibration Solution *10000X*
Leverandør: AAT BIOQUEST
67039
AATB67039EA 4925 NOK
AATB67039
MP650 Dye qPCR Calibration Solution *10000X*
Nukleinsyre reagenser qPCR/RT-qPCR Enzymes and Kits
MP650 is designed to have identical spectra to those of Mustang Purple.
  • Stable for 12 months
Quality tested in-house.
NB: For research use only.
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon