iFluor® 647, Succinimidyl Ester
Leverandør: AAT BIOQUEST
1031 71031 71510 71560
AATB1031EA 3600 NOK
AATB1031 AATB71031 AATB71510 AATB71560
iFluor® 647, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
NB: For research use only.