XFD750 NHS ester [Same Structure to Alexa Fluor™ 750 NHS ester]
Leverandør: AAT BIOQUEST
71912 1839
AATB71912EA 12875 NOK
AATB71912 AATB1839
XFD750 NHS ester [Same Structure to Alexa Fluor™ 750 NHS ester]
XFD750 NHS ester
XFD750 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 750 (Alexa Fluor® is the trademark of ThermoFisher).
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only