ReadiCleave™ AML Cleavage Buffer, 5X Aqueous Solution, AAT Bioquest
Leverandør: AAT BIOQUEST
7510
AATB7510EA 1525 NOK
AATB7510
ReadiCleave™ AML Cleavage Buffer, 5X Aqueous Solution, AAT Bioquest
Buffere Buffere, biologiske
ReadiCleave™ AML Cleavage Buffer provides an optimised formula to effectively remove the fluorescence tags from ReadiCleave™ AML-labeled biological targets when needed.
  • Designed for ease of use
  • Saves time
The removal of the fluorescent ReadiCleave™ AML tags may make the fluorescence imaging more compatible with the downstream analysis of a biological target by other techniques, e.g., MS analysis.
NB: For research use only.