Fluorescein aldehyde [5-FAM aldehyde] for biomolecular labeling
Leverandør: AAT BIOQUEST
Warning
9000
AATB9000EA 5150 NOK
AATB9000
Fluorescein aldehyde [5-FAM aldehyde] for biomolecular labeling
Fluorescein aldehyde [5-FAM aldehyde]
Fluorescein aldehyde is a reactive fluorescent dye that can react with an amine, hydrazine or hydroxylamine.
  • Reactive with amine, hydrazine, or hydroxylamine
  • Detectable with the FITC channel
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1d82bece87a9c2d566e6ae8c8ca320975354ce7&vid=b12daabc8abe2590f819ab6a1e1eb7f4cfe74bf0&prid=web&clid=SAPDAM