Fluorescein aldehyde [5-FAM aldehyde]
Leverandør: AAT BIOQUEST
9001
AATB9001EA 3325 NOK
AATB9001
Fluorescein aldehyde [5-FAM aldehyde]
Fluorescein aldehyde [5-FAM aldehyde]
Fluorescein aldehyde is a reactive fluorescent dye that can react with an amine, hydrazine or hydroxylamine.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only