Rhodamine aldehyde [5-TAMRA aldehyde] labeling_biomolecular

Leverandør: AAT BIOQUEST
Warning

9005
AATB9005EA 5150 NOK
AATB9005
Rhodamine aldehyde [5-TAMRA aldehyde] labeling_biomolecular
Rhodamine aldehyde [5-TAMRA aldehyde]

Rhodamine aldehyde is a reactive fluorescent dye that can react with an amine, hydrazine or hydroxylamine.


  • Reactive with amine, hydrazine, or hydroxylamine
  • Detectable with the FITC channel

MW: 536.03 g/mol
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58aee24fa530bc26fe674e7e1f1e81b28bf8032e&vid=a003beeb4346b231ecb246d8bf172ec44ca712c0&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR