Rhodamine aldehyde [5-TAMRA aldehyde]
Leverandør: AAT BIOQUEST
9006
AATB9006EA 3325 NOK
AATB9006
Rhodamine aldehyde [5-TAMRA aldehyde]
Rhodamine aldehyde [5-TAMRA aldehyde]
Rhodamine aldehyde is a reactive fluorescent dye that can react with an amine, hydrazine or hydroxylamine.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only