CytoCite™ Sample Tubes
Leverandør: AAT BIOQUEST
CCT100
AATBCCT100EA 1575 NOK
AATBCCT100
CytoCite™ Sample Tubes
Rør Reagensrør
Cytocite™ sample tubes are 500 μl thin-walled polypropylene tubes validated for use with Cytocite or Qubit Fluorometer.
  • Storage at room temperature
Optimised for easy use with both common fluorometer instruments.
NB: Research use only.