Anti-Biotin Rabbit Polyclonal Antibody (TRITC (Rhodamine))

Leverandør: Abcam
AB106226-100
ABCAAB106226-100EA 12075 NOK
ABCAAB106226-100
Anti-Biotin Rabbit Polyclonal Antibody (TRITC (Rhodamine))
Antistoffer
Type: Primary
Antigen: Biotin (biotin)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR